L-Lysinamide dihydrochloride, white powder

Supplier: MP Biomedicals

0215528780 0215528783 0215528791
ICNA0215528780EA 64.7 GBP
ICNA0215528780 ICNA0215528783 ICNA0215528791
L-Lysinamide dihydrochloride, white powder
L-Lysinamide dihydrochloride

Soluble in a 5% aqueous solution (clear, colorless solution).

Formula: C₆H₁₇Cl₂N₃O
MDL Number: MFCD00058287
CAS Number: 51127-08-3

Order Now

Specification Test Results

Form White Powder
Storage Store at -0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR