Oxalic acid dihydrate ≥99.5%, crystals ACS

Supplier: MP Biomedicals

0215255280 0215255290
ICNA0215255280EA 35 GBP
ICNA0215255280 ICNA0215255290
Oxalic acid dihydrate ≥99.5%, crystals ACS
Oxalic acid dihydrate

Insoluble in water.


An inner salt

Formula: HO₂CCO₂H·2H₂O
MDL Number: MFCD00149102
CAS Number: 6153-56-6
UN: 3261
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dcc1447f5f23a589dc85e10dd090c86e33b6541b&vid=2f0e34543b2f625715b002905fc0dd45fcd75561&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Synonyms Ethanedioic Acid Dihydrate
Form White Crystals
Storage Store at Room Temperature (15 to 30 °C)
Assay ≥99,5%
Heavy Metals (as Pb) ≤5 ppm
Insoluble Matter ≤0,005%
Residue on Ignition ≤0,01%
Chloride ≤0,002%
Sulphate ≤0,005%
Calcium ≤0,001%
Nitrogen Compounds (as N) ≤0,001%
Iron ≤2 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR