5-Acetylvaleric acid 90+%, Technical Grade

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 6-Oxoheptanoic acid

449530050 449530250 449531000
ACRO449530050EA 78.7 GBP
ACRO449530050 ACRO449530250 ACRO449531000
5-Acetylvaleric acid 90+%, Technical Grade
5-Acetylvaleric acid
Formula: C₇H₁₂O₃
Boiling Pt: 138…140 °C (1,5 mmHg)
Melting Pt: 35…37 °C
Storage Temperature: Ambient
MDL Number: MFCD00051479
CAS Number: 3128-07-2
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance White to off-white Powder or crystals
Infrared spectrum Conforms
GC ≥90.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR