Gold trichloride 99.99+% Trace metals basis
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
Gold(III) chloride, Gold (III) chloride
437080010
437080050
ACRO437080010EA
492
GBP
ACRO437080010
ACRO437080050
Gold trichloride 99.99+% Trace metals basis
Gold trichloride
Formula:
AuCl₃ MW: 303.32 g/mol Melting Pt: 254 °C Density: 4.3 Storage Temperature: Ambient |
MDL Number:
MFCD00014172 CAS Number: 13453-07-1 Merck Index: 13,04536 |
Specification Test Results
Appearance | Red to brown Crystalline powder or crystals |
Assay | 63.0 to 66.9 % (Au) |
Total trace metal impurities | ≤100 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...