2-Methyl-1-propenylmagnesium bromide 0.5 M THF, AcroSeal®

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 2-methyl-1-propenyl magnesium bromide

435568000 435561000
435568000.EA 1170 GBP
435568000. 435561000.
2-Methyl-1-propenylmagnesium bromide 0.5 M THF, AcroSeal®
2-Methyl-1-propenylmagnesium bromide
Formula: C₄H₇BrMg
MW: 159.31 g/mol
Density: 0.952 g/cm³
Flash Pt: –15 °C
Storage Temperature: Ambient
MDL Number: MFCD00209600
CAS Number: 38614-36-7
UN: 3399
ADR: 4.3 I

Order Now

Specification Test Results

Appearance Yellow to orange Solution
Titration Grignard 0.45 to 0.65 M
Refractive index 1.3390 to 1.3410 (20°C, 589 nm)
GC ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR