Dichlorocyclohexylmethylsilane 97+%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
Cyclohexylmethyldichlorosilane
432220100
432220500
ACRO432220100EA
95.5
GBP
ACRO432220100
ACRO432220500
Dichlorocyclohexylmethylsilane 97+%
Dichlorocyclohexylmethylsilane
Formula:
C₇H₁₄Cl₂Si MW: 197,17 g/mol Boiling Pt: 202 °C Density: 1,095 g/cm³ Flash Pt: 66 °C Storage Temperature: Ambient |
MDL Number:
MFCD00046333 CAS Number: 5578-42-7 UN: 2987 ADR: 8,II |
Specification Test Results
Appearance | Clear colorless to light brown Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4710 to 1.4730 (20°C, 589 nm) |
GC | ≥97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...