4-Vinylbenzyl acetate 95% stabilised

Supplier: Thermo Scientific
Warning

Synonyms: (4-ethenylphenyl)methyl acetate, (4-vinylphenyl)methyl acetate, 4-acetoxymethylstyrene

422500050 422500250
ACRO422500050EA 151 GBP
ACRO422500050 ACRO422500250
4-Vinylbenzyl acetate 95% stabilised
4-Vinylbenzyl acetate
Formula: C₁₁H₁₂O₂
MW: 176,215 g/mol
Boiling Pt: 125 °C (12 mmHg)
Density: 1,53 g/cm³
Flash Pt: >93 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00078265
CAS Number: 1592-12-7

Order Now

Specification Test Results

Appearance Clear light yellow to light pink Liquid
Infrared spectrum Conforms
Refractive index 1.5355 to 1.5385 (20°C, 589 nm)
GC ≥94 %
Stabilizer 0.1 % hydroquinone

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR