3-Nitrosalicylic acid 98%

Supplier: Thermo Scientific
Warning

Synonyms: 3-Nitrosalicylic Acid, 2-Hydroxy-3-nitrobenzoic acid

399820010 399820050
ACRO399820010EA 54.3 GBP
ACRO399820010 ACRO399820050
3-Nitrosalicylic acid 98%
3-Nitrosalicylic acid
Formula: C₇H₅NO₅
MW: 183,1183 g/mol
Melting Pt: 142…147 °C
Storage Temperature: Ambient
MDL Number: MFCD00024240
CAS Number: 85-38-1

Order Now

Specification Test Results

Appearance Yellow Crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR