Ethanol denatured 95+% (denatured with 5% methanol)

Supplier: Thermo Scientific
Danger

Synonyms: Ethyl alcohol denatured

365780010 365780025 365780100
ACRO365780010EA 75.1 GBP
ACRO365780010 ACRO365780025 ACRO365780100
Ethanol denatured 95+% (denatured with 5% methanol)
Ethanol denatured
Formula: H₃CCH₂OH
MW: 46,07 g/mol
Boiling Pt: ∼78 °C (1013 hPa)
Melting Pt: –117 °C
Density: 0,8 g/cm³
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00003568
CAS Number: 64-17-5
UN: 1170
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Refractive index 1.3600 to 1.3630 (20°C, 589 nm)
Density (20°C) 0.780 to 0.807 kg/l
GC ≤5 % (wood spirit)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR