(S)-2-(Aminomethyl)pyrrolidine 98%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
(S)-(+)-2-(Aminomethyl)pyrrolidine
348382500
348380010
ACRO348382500EA
142
GBP
ACRO348382500
ACRO348380010
(S)-2-(Aminomethyl)pyrrolidine 98%
(S)-2-(Aminomethyl)pyrrolidine
Formula:
C₅H₁₂N₂ MW: 100,17 g/mol Boiling Pt: 65 °C (11 mmHg) Density: 0,933 g/cm³ Flash Pt: 47 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00191745 CAS Number: 69500-64-7 UN: 2734 ADR: 8,II |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
GC | ≥97.5 % |
Specific optical rotation | +19.5° to +20.5° (20°C, 589 nm) (C=1, Chloroform) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...