Fluoxymesterone 97,0-102,0

Supplier: Thermo Fisher Scientific
Warning

124590010 124590050
ACRO124590010EA 927 GBP
ACRO124590010 ACRO124590050
Fluoxymesterone 97,0-102,0
Fluoxymesterone
Formula: C₂₀H₂₉FO₃
MW: 336.45 g/mol
Melting Pt: 240 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00010480
CAS Number: 76-43-7

Order Now

Specification Test Results

Appearance White to off-white Powder (May discolor to light yellow (Photosensitive))
Infrared spectrum Conforms
Loss on drying =1 % (105°C, 3 h)
HPLC 97.0 to 102.0 % (on dried substance)
Specific optical rotation +104° to +112° (20°C, 589 nm) (c=1, EtOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR