Dimethyl methylphosphonate 97%

Supplier: Thermo Fisher Scientific
Danger

116420050 116420250
ACRO116420050EA 43.6 GBP
ACRO116420050 ACRO116420250
Dimethyl methylphosphonate 97%
Dimethyl methylphosphonate
Formula: C₃H₉O₃P
MW: 124.08 g/mol
Boiling Pt: 181 °C (760 mmHg)
Density: 1.14 g/cm³
Flash Pt: 68 °C
Storage Temperature: Ambient
MDL Number: MFCD00008349
CAS Number: 756-79-6
Merck Index: 13,03425

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4120 to 1.4140 (20°C, 589 nm)
GC ≥96.0 %
Sulfate (SO4) ≤0.002 %
Iron (Fe) ≤2 ppm
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.005 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR