4,4,5,5-Tetramethyl-2-vinyl-1,3,2-dioxaborolane 97+% stabilised
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
Pinacol vinylboronate, Vinylboronic Acid Pinacol Ester
Stabilisation:
0.05% Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
395530010
395530050
ACRO395530010EA
106
GBP
ACRO395530010
ACRO395530050
4,4,5,5-Tetramethyl-2-vinyl-1,3,2-dioxaborolane 97+% stabilised
4,4,5,5-Tetramethyl-2-vinyl-1,3,2-dioxaborolane
Appearance: Clear colorless to amber Liquid
Formula:
C₈H₁₅BO₂ MW: 154.02 g/mol Boiling Pt: 40 °C (20 mmHg) Density: 0.908 g/cm³ Flash Pt: 34 °C Storage Temperature: Freezer |
MDL Number:
MFCD00192492 CAS Number: 75927-49-0 UN: 3272 ADR: 3 III |
Specification Test Results
Appearance | Clear colorless to amber Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4240 to 1.4260 (20°C, 589 nm) |
GC | ≥97.0 % |
Stabilizer | ≤500 ppm BHT |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...