3-Ethynyltoluene 97%

Supplier: Thermo Fisher Scientific
Danger

392660010 392660050
ACRO392660010EA 66.6 GBP
ACRO392660010 ACRO392660050
3-Ethynyltoluene 97%
3-Ethynyltoluene
Formula: C₉H₈
Flash Pt: 25 °C
Storage Temperature: Ambient
MDL Number: MFCD01318174
CAS Number: 766-82-5
UN: 3295
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow to brown Liquid
Infrared spectrum Conforms
Refractive index 1.5400 to 1.5450 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR