3,3-Dimethylbutylamine 95%

Supplier: Thermo Fisher Scientific
Danger

179380010 179380050
ACRO179380010EA 162 GBP
ACRO179380010 ACRO179380050
3,3-Dimethylbutylamine 95%
3,3-Dimethylbutylamine

Appearance: Clear colorless to yellow Liquid

Formula: C₆H₁₅N
MW: 101.193 g/mol
Boiling Pt: 114…116 °C (760 mmHg)
Density: 0.752 g/cm³
Flash Pt: 6 °C
Storage Temperature: Ambient
MDL Number: MFCD00008201
CAS Number: 15673-00-4
UN: 3286
ADR: 3 II

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4120 to 1.4160 (20°C, 589 nm)
GC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR