Ethynylferrocene 97%

Supplier: Thermo Scientific
Warning

Synonyms: (2α)-EthynylferroceneFerrocene, ethynyl-, (2α)-1-cyclopenta-2,4-dienyl-(2-ethynyl-1-cyclopenta-2,4-dienyl)iron

ACRO378830010EA 241 GBP
ACRO378830010
Ethynylferrocene 97%
Ethynylferrocene

Appearance: Red-brown Crystalline powder or crystals

Formula: C₁₂H₁₀Fe
MW: 210,057 g/mol
Melting Pt: 50…57 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03701501
CAS Number: 1271-47-2

Order Now

Specification Test Results

Appearance Red-brown Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 50 to 57 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR