Dimethyl aminomalonate hydrochloride 97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Dimethyl 2-aminomalonate hydrochloride
374970050
374970250
ACRO374970050EA
149
GBP
ACRO374970050
ACRO374970250
Dimethyl aminomalonate hydrochloride 97%
Dimethyl aminomalonate hydrochloride
Appearance: Light yellow Crystalline powder or crystals
Formula:
C₅H₁₀ClNO₄ MW: 183,588 g/mol Melting Pt: 155 °C Storage Temperature: Ambient |
MDL Number:
MFCD00038880 CAS Number: 16115-80-3 |
Specification Test Results
Appearance | Light yellow Crystalline powder or crystals |
Infrared spectrum | Conforms |
Titration Argentometric | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...