Xanthotoxin 99%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 9-Methoxy-7H-furo[3,2-g]chromen-7-one, 8-Methoxypsoralen, Methoxsalen

214150010 214150050
ACRO214150010EA 95.8 GBP
ACRO214150010 ACRO214150050
Xanthotoxin 99%
Xanthotoxin

Appearance: White to cream Powder or crystals or needles

Formula: C₁₂H₈O₄
MW: 216.19 g/mol
Melting Pt: 143…148 °C
Storage Temperature: Ambient
MDL Number: MFCD00005009
CAS Number: 298-81-7
UN: 3261
ADR: 8 II

Order Now

Specification Test Results

Appearance White to cream Powder or crystals or needles
Infrared spectrum Conforms
Melting point 143 to 148 °C
Water ≤0.5 % (K.F.)
HPLC ≥98.5 % (on anhydrous substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR