(±)-2-Phenylpropanal 98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
DL-2-Phenylpropionaldehyde, (±)-2-Phenylpropionaldehyde, (±)-Hydratropaldehyde
130860250
130861000
130865000
ACRO130860250EA
42.2
GBP
ACRO130860250
ACRO130861000
ACRO130865000
(±)-2-Phenylpropanal 98%
(±)-2-Phenylpropanal
Formula:
C₉H₁₀O MW: 134,18 g/mol Boiling Pt: 202…205 °C (760 mmHg) Melting Pt: –60 °C Density: 1,011 g/cm³ Flash Pt: 76 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006973 CAS Number: 93-53-8 |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5160 to 1.5240 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...