Cinnamamide predominantly trans 97%

Supplier: Thermo Fisher Scientific
Warning

110360050 110360250
ACRO110360050EA 87.2 GBP
ACRO110360050 ACRO110360250
Cinnamamide predominantly trans 97%
Cinnamamide

CAS No.: 621-79-4

Formula: C₉H₉NO
Melting Pt: 146…150 °C
Storage Temperature: Ambient
MDL Number: MFCD00008033
CAS Number: 621-79-4

Order Now

Specification Test Results

Appearance White to beige Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 146 to 150 °C
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR