N-Carbobenzoxy-L-proline 99%

Supplier: Thermo Scientific

Synonyms: Z-Pro-OH, Z-L-Prolin, N-Cbz-L-proline, N-Carbobenzyloxy-L-proline, N-Benzyloxycarbonyl-L-proline

108370100 108370050
ACRO108370100EA 177 GBP
ACRO108370100 ACRO108370050
N-Carbobenzoxy-L-proline 99%
N-Carbobenzoxy-L-proline

CAS No.: 1148-11-4

Formula: C₁₃H₁₅NO₄
MW: 249,27 g/mol
Melting Pt: 76…78 °C
Storage Temperature: Ambient
MDL Number: MFCD00003170
CAS Number: 1148-11-4

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder or crystals
Infrared spectrum Conforms
Water ≤0.5 % (K.F.)
HPLC ≥98.5 %
Specific optical rotation -59° to -61° (20°C, 589 nm) (c=2, CH3COOH)
Titration with NaOH ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR