4-(Methylsulphonyl)phenylhydrazine hydrochloride 95%

Supplier: Thermo Scientific

Synonyms: (4-(Methylsulphonyl)phenyl)hydrazine hydrochloride

424960025 424960100
ACRO424960025EA 177 GBP
ACRO424960025 ACRO424960100
4-(Methylsulphonyl)phenylhydrazine hydrochloride 95%
4-(Methylsulphonyl)phenylhydrazine hydrochloride

Appearance: Off-white to light yellow Crystalline powder

Formula: C₇H₁₁ClN₂O₂S
MW: 222,687 g/mol
Melting Pt: 202 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00216494
CAS Number: 17852-67-4

Order Now

Specification Test Results

Appearance Off-white to light yellow Crystalline powder
Infrared spectrum Conforms
Titration Argentometric ≥94 %
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR