Glycogen (from oysters)

Supplier: Thermo Fisher Scientific

422950010 422950050
ACRO422950010EA 64.9 GBP
ACRO422950010 ACRO422950050
Glycogen (from oysters)
Glycogen

Appearance: White to light beige Powder

Formula: C₂₄H₄₂O₂₁
Storage Temperature: Refrigerator
MDL Number: MFCD00081547
CAS Number: 9005-79-2

Order Now

Specification Test Results

Appearance White to light beige Powder
Infrared spectrum Conforms
Loss on drying ≤10 % (105°C, 3 h)
Specific optical rotation +160.0° to +180.0° (20°C, 589 nm) (c=0.1, water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR