(3S,4S)-(+)-1-Benzyl-3,4-pyrrolidindiol 97%

Supplier: Thermo Scientific
Warning

Synonyms: (3S,4S)-1-Benzyl-3,4-pyrrolidindiol

348730010 348730050
ACRO348730010EA 229 GBP
ACRO348730010 ACRO348730050
(3S,4S)-(+)-1-Benzyl-3,4-pyrrolidindiol 97%
(3S,4S)-(+)-1-Benzyl-3,4-pyrrolidindiol

CAS No.: 90365-74-5

Formula: C₁₁H₁₅NO₂
MW: 193,2423 g/mol
Melting Pt: 95…101 °C
Storage Temperature: Ambient
MDL Number: MFCD01073893
CAS Number: 90365-74-5

Order Now

Specification Test Results

Appearance Off-white to yellow to light brown Powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 95 to 101 °C
GC ≥96.0 %
Specific optical rotation +29° to +35° (20 °C, 589 nm) (c=1, MeOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR