tert-Butyl (3S)-3-aminobutanoate 95%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
(S)-tert-Butyl 3-aminobutanoate
337941000
337940010
ACRO337941000EA
154
GBP
ACRO337941000
ACRO337940010
tert-Butyl (3S)-3-aminobutanoate 95%
tert-Butyl (3S)-3-aminobutanoate
CAS No.: 161105-54-0
Formula:
C₈H₁₇NO₂ MW: 159,229 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00798294 CAS Number: 161105-54-0 UN: 2735 ADR: 8,III |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4230 to 1.4270 (20°C, 589 nm) |
GC | ≥94 % |
Specific optical rotation | +21° to +27° (20°C, 589 nm) (c=1, CHCl3) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...