Zeolite Type: ZSM-5

Supplier: Thermo Scientific
Warning

279571000 279570050 279575000
ACRO279571000EA 148 GBP
ACRO279571000 ACRO279570050 ACRO279575000
Zeolite Type: ZSM-5
Zeolite

CAS No.: 308081-08-5

Formula: Na₂O·Al₂O₃·xSiO₂·yH₂O
Storage Temperature: Ambient
MDL Number: MFCD00132601
CAS Number: 308081-08-5

Order Now

Specification Test Results

Appearance White to beige to light brown Powder
Surface area ≥300 m²/g (BET)
Composition 400 to 570 Molar ratio SiO2/Al2O3
Sodium (Na) ≤700 ppm
Loss on ignition ≤10 % (1000°C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR