Zinc-copper couple

Supplier: Thermo Scientific
Warning

271440250 271441000
ACRO271440250EA 117 GBP
ACRO271440250 ACRO271441000
Zinc-copper couple
Zinc-copper couple

Appearance: Grey-black Powder

Formula: CuZn
MW: 128,926 g/mol
Melting Pt: 500 °C
Storage Temperature: Ambient
MDL Number: MFCD00084851
CAS Number: 53801-63-1
UN: 3089
ADR: 4.1,III

Order Now

Specification Test Results

Appearance Grey to black Powder
Copper (Cu) ≤2 %
Zinc (Zn) ≥98 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR