2-Acetamido-6-hydroxypurine 99%

Supplier: Thermo Fisher Scientific

Synonyms: N-Acetylguanine

258400010 258400050
ACRO258400010EA 95 GBP
ACRO258400010 ACRO258400050
2-Acetamido-6-hydroxypurine 99%
2-Acetamido-6-hydroxypurine

CAS No.: 19962-37-9

Formula: C₇H₇N₅O₂
MW: 193.17 g/mol
Melting Pt: >260 °C
Storage Temperature: Ambient
MDL Number: MFCD00078201
CAS Number: 19962-37-9

Order Now

Specification Test Results

Appearance White to beige Powder
Infrared spectrum Conforms
Titration with HClO4 ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR