3-Hydroxybenzylhydrazine dihydrochloride 98%

Supplier: Thermo Fisher Scientific
Warning

229030010 229030050
ACRO229030010EA 198 GBP
ACRO229030010 ACRO229030050
3-Hydroxybenzylhydrazine dihydrochloride 98%
3-Hydroxybenzylhydrazine dihydrochloride

Appearance: Off-white to beige Adhering powder

Formula: C₇H₁₂Cl₂N₂O
Melting Pt: 138 °C
Storage Temperature: Ambient
MDL Number: MFCD00044695
CAS Number: 81012-99-9

Order Now

Specification Test Results

Appearance Off-white to beige Adhering powder
Infrared spectrum Conforms
Titration Argentometric ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR