Anti-IgG Goat Polyclonal Antibody (APC (Allophycocyanin))

Supplier: Biotium
20414-100UL 20414-500UL
20414-100ULEA 200 GBP
20414-100UL 20414-500UL
Anti-IgG Goat Polyclonal Antibody (APC (Allophycocyanin))
Antibodies
This product is prepared by labeling F(ab’)2 fragment of goat anti-mouse IgG (H+L) with Allophycocyanin (APC). APC belongs to the family of phycobiliproteins, which are derived from cyanobacteria and eukaryotic algae. APC has absorption maxima at 650 and emission at 660 nm. Because of the high fluorescence quantum yield (~0.68), APC is by far the brightest dye for the 650 nm wavelength region.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: APC
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR