Anti-IgG Goat Polyclonal Antibody (RPE (R-Phycoerythrin))

Supplier: Biotium

20358-500UL 20358-200UL
BTIU20358-500ULEA 485 GBP
BTIU20358-500UL BTIU20358-200UL
Anti-IgG Goat Polyclonal Antibody (RPE (R-Phycoerythrin))
Antibodies
This product is prepared by labeling F(ab’)2 fragment of goat anti-rabbit IgG (H+L) with R-phycoerythrin (R-PE). R-PE belongs to the family of phycobiliproteins, which are derived from cyanobacteria and eukaryotic algae. R-PE has absorption maxima at 496, 546, and 565 nm respectively, and emission at 578 nm. Because of the high extinction coefficient (~1,960,000) and high fluorescence quantum yield (~0.82), R-PE is by far the brightest dye for the 565 nm wavelength region.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: R-PE
Epitope:
Host: Goat
Isotype:
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR