Desalting chromatography cartridges, 7K MWCO, Zeba™
Supplier: Thermo Fisher Scientific
89934 89935
PIER89934EA 265 GBP
PIER89934 PIER89935
Desalting chromatography cartridges, 7K MWCO, Zeba™
Chromatography Columns
The Zeba™ Desalting chromatography cartridges, 7 kDa MWCO, contain a high-performance resin for desalting or buffer exchange applications. This resin is ideal for removing low molecular weight compounds including salts, fluorescent dyes, biotin and other small labeling reagents. Zeba™ Desalting Cartridges provide fast, easy and reproducible chromatographic separations and can be regenerated for multiple uses. They efficiently process samples from 50 to 1500 µl.
  • Low-binding resin maximises protein recovery
  • No fraction screening or waiting for protein to emerge by gravity-flow
  • No cumbersome column preparation or equilibration making it easy to use
  • Purification Target: Buffer Exchange, Protein
  • Column Type: Size-Exclusion, Proprietary Resin
Zeba™ Desalting cartridge fittings are convenient, ready to use devices compatible with the major automated liquid-chromatography systems or for manual syringe processing. The cartridges attach directly to ÄKTA™ or FPLC Systems without additional connectors. An accessory pack included with each product readily adapts cartridges for use with Luer syringe fittings or ¹/₁₆" tubing.
Similarly, Zeba Desalting plates (7K MWCO) are polypropylene devices that contain Zeba High-Performance Resin and provide excellent protein desalting performance, with high recovery of proteins >7000 Da in a centrifuge format for small sample volumes. The plates enable fast, consistent, well-to-well and plate-to-plate reproducibility for small-scale, high-throughput separations. Zeba 96-well spin desalting plates require no resin dispensing or hydration and provide the same high protein recovery as spin columns with at least 95% removal of salts and other small molecules. One plate can process 96 small sample volumes (20 to 100 µl) in just five minutes, offering rapid and exceptional protein desalting.
The Zeba Desalting Chromatography Cartridges contain Zeba High-Performance Resin for desalting or buffer exchange applications. This resin is ideal for removing low molecular weight compounds including salts, fluorescent dyes, biotin, and other small labeling reagents. Protein samples can be processed with unsurpassed protein recovery and 95% retention of salts and other small molecular contaminants (<1,000 Da). Zeba Desalting cartridges provide fast, easy, and reproducible chromatographic separations and can be regenerated for multiple uses.
Delivery information: Each column pack contains one accessory pack with one female Luer-Lok® adapter, one connector fitting, one column plug and five bottom caps.