Metolazone, powder
Supplier: MP Biomedicals
Synonyms:
Zaroxolyn, 7-Chloro-1,2,3,4-tetrahydro-2-methyl-3-(2-methylphenyl)-4-oxo-6-quinazolinesulphonamide
0215361725
0215361780
ICNA0215361725EA
363
GBP
ICNA0215361725
ICNA0215361780
Metolazone, powder
Metolazone
Product Description: Diuretic, anti-hypertensive
Biochem/physiol Actions: Inhibitor of thiazide-sensitive Na+-Cl- cotransporter; antihypertensive; moderate "loop" diuretic.
Melting Point: 245-260°C
Presentation: White Crystalline Powder
Solubility: DMSO: >5 mg/mL
Storage & Handling: Room Temperature
Formula:
C₁₆H₁₆ClN₃O₃S MW: 365,832 g/mol |
MDL Number:
MFCD00069304 CAS Number: 17560-51-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...