Acetoacetyl Coenzyme A Trisodium Salt Tetrahydrate ≥90%

Supplier: MP Biomedicals

0215022405 0215022401 0215022425 0215022450
ICNA0215022405EA 265 GBP
ICNA0215022405 ICNA0215022401 ICNA0215022425 ICNA0215022450
Acetoacetyl Coenzyme A Trisodium Salt Tetrahydrate ≥90%
Acetoacetyl coenzyme a trisodium salt tetrahydrate

Acetoacetyl-CoA is a precursor of poly-beta-hydroxybutyrate polymers in microorganisms. It plays an important role in the biosynthesis of nonactin.


It may be used as a substrate for a number of enzymes including acetoacetyl-CoA thiolase which produces acetyl-CoA in reverse condensation reaction and (3-Hydroxy-3-methylglutaryl coenzyme A (CoA) synthase (HMGCS)) which produces 3-hydroxy-3-methylglutaryl (HMG)-CoA in the mevalonate pathway leading to terpenoid biosynthesis.


Soluble in water (50 mg/ml - clear colorless to faint yellow solution).

Formula: C₂₅H₃₇N₇O₁₈P₃SNa₃·4H₂O
MW: 989.56 g/mol
CAS Number: 102029-52-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR