Potassium hydrogen carbonate, granular USP
Supplier: Spectrum Chemical
Synonyms:
Potassium bicarbonate
PO158-12KG
PO158-2.5KG
PO158-45KGBL
SPCMPO158-12KGEA
1170
GBP
SPCMPO158-12KG
SPCMPO158-2.5KG
SPCMPO158-45KGBL
Potassium hydrogen carbonate, granular USP
Potassium hydrogen carbonate
Kosher certified
BSE/TSE statement (available upon request)
Formula:
KHCO₃ MW: 100.12 g/mol Melting Pt: 292 Density: 2.17 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011402 CAS Number: 298-14-6 EINECS: 206-059-0 Merck Index: 13,07691 |
Specification Test Results
Assay | 99.5% Min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...