Ethyl-2-methyl-3-hydroxy-4,4,4-trifluorobutyrate 97%
Supplier: Thermo Scientific
|
Warning
|
L19969.03
L19969.06
L19969.03EA
41.6
GBP
L19969.03
L19969.06
Ethyl-2-methyl-3-hydroxy-4,4,4-trifluorobutyrate 97%
Ethyl-2-methyl-3-hydroxy-4,4,4-trifluorobutyrate
Purity: 97%
CAS No.: 91600-33-8
Molecular Formula: C7H11F3O3
Formula Weight: 200.16
Boiling Point: 180-184°
Beilstein Reference: 6194166
MDL No.: MFCD00190634
Formula:
C₇H₁₁F₃O₃ Boiling Pt: 180…184 °C Storage Temperature: Ambient |
MDL Number:
MFCD00190634 CAS Number: 91600-33-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...