4-sulphamidobenzoyl chloride DMF complex 95%

Supplier: Thermo Fisher Scientific
Danger

L18428.06 L18428.03
L18428.06EA 85.1 GBP
L18428.06 L18428.03
4-sulphamidobenzoyl chloride DMF complex 95%
4-sulphamidobenzoyl chloride DMF complex

Purity: 95% CAS No.: 1172493-99-0 Molecular Formula: C10H13ClN2O4S Formula Weight: 292.74 UN No.: UN3261 MDL No.: MFCD02094019

Formula: C₁₀H₁₃ClN₂O₄S
MW: 292.74 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02094019
CAS Number: 1172493-99-0
UN: 3261
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR