5-(3-Phenoxyphenyl)-1H-tetrazole 98%

Supplier: Thermo Scientific
Warning

L17955.03 L17955.MD
L17955.03EA 0 GBP
L17955.03 L17955.MD
5-(3-Phenoxyphenyl)-1H-tetrazole 98%
5-(3-Phenoxyphenyl)-1H-tetrazole

Purity: 98% CAS No.: 374538-02-0 Molecular Formula: C13H10N4O Formula Weight: 238.25 Melting Point: 152-154° MDL No.: MFCD02093975

Formula: C₁₃H₁₀N₄O
MW: 238,25 g/mol
Melting Pt: 152…154 °C
Storage Temperature: Ambient
MDL Number: MFCD02093975
CAS Number: 374538-02-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR