3-Amino-4-methylphenylboronic acid 98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
3-Amino-4-methylbenzeneboronic Acid, 3-Amino-p-tolylboronic Acid, 3-Amino-4-methylphenylboronic Acid
L17695.06
L17695.03
L17695.06EA
165
GBP
L17695.06
L17695.03
3-Amino-4-methylphenylboronic acid 98%
3-Amino-4-methylphenylboronic acid
Purity: 98%
CAS No.: 22237-12-3
Molecular Formula: C7H10BNO2
Formula Weight: 150.98
Melting Point: ca 250°
MDL No.: MFCD01074640
Formula:
C₇H₁₀BNO₂ MW: 150,97 g/mol Melting Pt: ∼ 250 °C Storage Temperature: Ambient |
MDL Number:
MFCD01074640 CAS Number: 22237-12-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...