(Diphenylphosphonimido)triphenylphosphorane ≥98%

Supplier: Thermo Scientific

L17582.14 L17582.06
L17582.14EA 186 GBP
L17582.14 L17582.06
(Diphenylphosphonimido)triphenylphosphorane ≥98%
(Diphenylphosphonimido)triphenylphosphorane

Purity: 98% CAS No.: 2156-69-6 Molecular Formula: C30H25NOP2 Formula Weight: 477.49 Melting Point: 170-174° MDL No.: MFCD00045685

Formula: C₃₀H₂₅NOP₂
MW: 477,49 g/mol
Melting Pt: 170…174 °C
Storage Temperature: Ambient
MDL Number: MFCD00045685
CAS Number: 2156-69-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR