1,8-Diazabicyclo[5.4.0]-7-undecene hydrogen tribromide 98%

Supplier: Thermo Scientific
Danger

Synonyms: 2,3,4,6,7,8,9,10-Octahydropyrimido[1,2-a]azepine Hydrogen Tribromide, 1,8-Diazabicyclo[5.4.0]-7-undecene Hydrogen Tribromide

L17569.03 L17569.06
L17569.03EA 28.4 GBP
L17569.03 L17569.06
1,8-Diazabicyclo[5.4.0]-7-undecene hydrogen tribromide 98%
1,8-Diazabicyclo[5.4.0]-7-undecene hydrogen tribromide

Purity: 98% CAS No.: 138666-59-8 Molecular Formula: C9H17Br3N2 Formula Weight: 392.96 Melting Point: 119-122° UN No.: UN3261 MDL No.: MFCD00191982

Formula: C₉H₁₆N₂
MW: 392,96 g/mol
Melting Pt: 119…122 °C
Storage Temperature: Ambient
MDL Number: MFCD00191982
CAS Number: 138666-59-8
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR