2,4,6-Tris(pentadecafluoroheptyl)-1,3,5-triazine mass spec standard

Supplier: Thermo Scientific

Synonyms: 2,4,6-Tris(perfluoroheptyl)-1,3,5-triazine

L16678.ME
L16678.MEEA 99 GBP
L16678.ME
2,4,6-Tris(pentadecafluoroheptyl)-1,3,5-triazine mass spec standard
2,4,6-Tris(pentadecafluoroheptyl)-1,3,5-triazine

CAS No.: 21674-38-4 Molecular Formula: C24F45N3 Formula Weight: 1185.22 Melting Point: 29-31° Boiling Point: 150°/1.5mm MDL No.: MFCD00042439

Formula: C₂₄F₄₅N₃
MW: 1185,21 g/mol
Boiling Pt: 150 °C (1,5 mmHg)
Melting Pt: 29…31 °C
Storage Temperature: Ambient
MDL Number: MFCD00042439
CAS Number: 21674-38-4
EINECS: 244-521-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR