O,O'-Bis(trimethylsilyl)-5-fluorouracil 97%

Supplier: Thermo Scientific
Danger

Synonyms: 5-Fluoro-2,4-bis((trimethylsilyl)oxy)pyrimidine

L16407.06 L16407.14
L16407.06EA 78.6 GBP
L16407.06 L16407.14
O,O'-Bis(trimethylsilyl)-5-fluorouracil 97%
O,O'-Bis(trimethylsilyl)-5-fluorouracil

Purity: 97% CAS No.: 17242-85-2 Molecular Formula: C10H19FN2O2Si2 Formula Weight: 274.44 Flash Point: 32°(89°F) Refractive Index: 1.4590 Boiling Point: 114°/14mm Beilstein Reference: 885268 UN No.: UN1992 MDL No.: MFCD00042441

Formula: C₁₀H₁₉FN₂O₂Si₂
MW: 274,44 g/mol
Boiling Pt: 114 °C (14 mmHg)
Density: 1,050
Flash Pt: 32 °C (89 °F)
Storage Temperature: Ambient
MDL Number: MFCD00042441
CAS Number: 17242-85-2
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR