(+)-Menthyl acetate 99%

Supplier: Thermo Scientific
Warning

Synonyms: (+)-Acetoxy-p-menthane

L13987.03 L13987.MD
L13987.03EA 68.2 GBP
L13987.03 L13987.MD
(+)-Menthyl acetate 99%
(+)-Menthyl acetate

Purity: 99% CAS No.: 5157-89-1 Molecular Formula: C12H22O2 Formula Weight: 198.31 Flash Point: 77°(170°F) Refractive Index: 1.4470 Boiling Point: 229-230° Beilstein Reference: 1909419 Optical Rotation: +80.5° (c=8 in benzene) MDL No.: MFCD00062977

Formula: C₁₂H₂₂O₂
MW: 198,31 g/mol
Boiling Pt: 229…230 °C
Density: 0,925
Flash Pt: 77 °C (170 °F)
Storage Temperature: Ambient
MDL Number: MFCD00062977
CAS Number: 5157-89-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR