1-Chloro-1,1-difluoro-2,4-pentanedione ≥97%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
1-Chloro-1,1-difluoroacetylacetone
L13027.06
L13027.14
L13027.06EA
0
GBP
L13027.06
L13027.14
1-Chloro-1,1-difluoro-2,4-pentanedione ≥97%
1-Chloro-1,1-difluoro-2,4-pentanedione
Purity: 97%
CAS No.: 2375-76-0
Molecular Formula: C5H5ClF2O2
Formula Weight: 170.54
Refractive Index: 1.4345
Boiling Point: 141°
Beilstein Reference: 1858592
UN No.: UN1993
MDL No.: MFCD00041520
Formula:
C₅H₅ClF₂O₂ Boiling Pt: 141 °C Storage Temperature: Ambient |
MDL Number:
MFCD00041520 CAS Number: 2375-76-0 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...