Cyclopropylacetonitrile 97%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 2-Cyclopropylacetonitrile

L11950.06 L11950.14
L11950.06EA 72.4 GBP
L11950.06 L11950.14
Cyclopropylacetonitrile 97%
Cyclopropylacetonitrile

Purity: 97% CAS No.: 6542-60-5 Molecular Formula: C5H7N Formula Weight: 81.12 Flash Point: 43°(109°F) Refractive Index: 1.4245 Odour: Strong Boiling Point: 142-144° Beilstein Reference: 2036964 UN No.: UN3275 MDL No.: MFCD00041523

Formula: C₅H₇N
MW: 81.12 g/mol
Boiling Pt: 142…144 °C
Density: 0.890
Flash Pt: 43 °C (109 °F)
Storage Temperature: Ambient
MDL Number: MFCD00041523
CAS Number: 6542-60-5
ADR: 6.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR