tert-Butyl (1S)-1-(hydroxymethyl)-2-methylpropylcarbamate ≥97%

Supplier: Thermo Fisher Scientific

Synonyms: N-BOC-L-Valinol, Boc-Valinol, (S)-tert-butyl (1-hydroxy-3-methylbutan-2-yl)carbamate

L10236.03 L10236.MD
L10236.03EA 0 GBP
L10236.03 L10236.MD
tert-Butyl (1S)-1-(hydroxymethyl)-2-methylpropylcarbamate ≥97%
tert-Butyl (1S)-1-(hydroxymethyl)-2-methylpropylcarbamate

Purity: 97% CAS No.: 79069-14-0 Molecular Formula: C10H21NO3 Formula Weight: 203.28 Storage Temperature: 2°C to 8°C Flash Point: 85°(185°F) Refractive Index: 1.4510 Boiling Point: 218° Beilstein Reference: 4663728 Optical Rotation: +22° (c=1 in chloroform) MDL No.: MFCD00082635

Formula: C₁₀H₂₁NO₃
MW: 203.28 g/mol
Boiling Pt: 218 °C
Flash Pt: 85 °C (185 °F)
MDL Number: MFCD00082635
CAS Number: 79069-14-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR