(3S)-(+)-1-Benzyl-3-aminopyrrolidine 99%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
(S)-1-Benzyl-3-aminopyrrolidine, (S)-(+)-1-Benzyl-3-aminopyrrolidine
L09449.03
L09449.06
L09449.03EA
87.3
GBP
L09449.03
L09449.06
(3S)-(+)-1-Benzyl-3-aminopyrrolidine 99%
(3S)-(+)-1-Benzyl-3-aminopyrrolidine
Purity: 99%
CAS No.: 114715-38-7
Molecular Formula: C11H16N2
Formula Weight: 176.26
Flash Point: 140°(284°F)
Refractive Index: 1.5440
Boiling Point: 100-105°/3mm
Beilstein Reference: 6477593
Optical Rotation: +5° (c=10 in ethanol)
UN No.: UN2735
MDL No.: MFCD00082766
Formula:
C₁₁H₁₆N₂ MW: 176,26 g/mol Boiling Pt: 100…105 °C (3 mmHg) Density: 1,020 Flash Pt: 140 °C (284 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00082766 CAS Number: 114715-38-7 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...