N,N-Dimethyldecanamide 98%
Supplier: Thermo Fisher Scientific
Synonyms:
N,N-Dimethylcapramide
L08731.06
L08731.14
L08731.06EA
80.9
GBP
L08731.06
L08731.14
N,N-Dimethyldecanamide 98%
N,N-Dimethyldecanamide
Purity: 98%
CAS No.: 14433-76-2
Molecular Formula: CH3(CH2)8CON(CH3)2
Formula Weight: 199.34
Refractive Index: 1.4540
Boiling Point: 110-111°/0.5mm
Beilstein Reference: 1906042
MDL No.: MFCD00043725
Formula:
C₁₂H₂₅NO MW: 199.34 g/mol Boiling Pt: 110…111 °C (0.5 mmHg) Storage Temperature: Ambient |
MDL Number:
MFCD00043725 CAS Number: 14433-76-2 EINECS: 238-405-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...