Methyl methanesulphonylacetate ≥98%

Supplier: Thermo Fisher Scientific

Synonyms: Methanesulphonylacetic acid Methyl Ester, Methyl 2-(methylsulphonyl)acetate

L08039.14 L08039.06
L08039.14EA 323 GBP
L08039.14 L08039.06
Methyl methanesulphonylacetate ≥98%
Methyl methanesulphonylacetate

Purity: 98+% CAS No.: 62020-09-1 Molecular Formula: C4H8O4S Formula Weight: 152.17 Melting Point: 59-63° Beilstein Reference: 2412782 MDL No.: MFCD00051842

Formula: C₄H₈O₄S
MW: 152.16 g/mol
Melting Pt: 59…63 °C
Storage Temperature: Ambient
MDL Number: MFCD00051842
CAS Number: 62020-09-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR